Orders LinkContactUsLinkComponent
Ab buffer kit
Ab buffer kit
Ab Buffer Kit, 1 KIT
Catalog # 28-9030-59
Supplier:   Cytiva
Ab buffer kit
Catalog # 28-9030-59
Supplier:  Cytiva
CAS Number:  
Ab buffer kit
Ab buffer kit
Ab Buffer Kit, 1 KIT
CAS Number:  
Price
179,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents
Ab buffer kit contains prepared buffer concentrates for binding, washing, and elution of IgG according to recommended protocols.
The kit also includes neutralising buffer. Ab buffer kit eliminates time consuming buffer preparation, which allows fast, reproducible, and convenient purification work.
Delivery information: Includes: 1×50 ml binding buffer (10X stock solution), 1×15 ml elution buffer (10X stock solution), and 1×25 ml neutralising buffer.
Specifications
Specifications
  • VPE:
    1 KIT
  • Omschrijving:
    Ab Buffer Kit
Specifications