Waste bin, B2, Tork®
wit, 20 L, 216×326×377 mm
Catalogus # 115-1621
Leverancier: Essity
Ga naar:
Product Details & Documents
Plastic.
- Practical
- Easy to use
Wall-mounted small model.
Specificaties
- Capaciteit:20 L
- Kleur:wit
- b×d×h:216×326×377 mm