Universal drum tops
Universal drum top, for 200 l drum, Ø 560 mm, 25 tops/pack
Catalogus # MSGA9953
Leverancier: asecos
Ga naar:
Product Details & Documents
Universal absorbent mat for drum tops for safe absorption of oil, water, cooling liquids and non aggressive chemicals.
- Suitable for 200 litre drums
- Optimal protection while dispensing ensuring drum top and floor stay clean
- Absorbent capacity up to 16 times its own weight
Specificaties
- Omschrijving:Universal drum top, for 200 l drum, Ø 560 mm
- Verpakt:25 tops/pack