Orders LinkContactUsLinkComponent
Affinity chromatography columns, Ab SpinTrap™
Affinity chromatography columns, Ab SpinTrap™
Ab SpinTrap™, 34 µm, 100 µl
Catalogus # 28-4083-47
Leverancier:   Cytiva
Affinity chromatography columns, Ab SpinTrap™
Catalogus # 28-4083-47
Leverancier:  Cytiva
Affinity chromatography columns, Ab SpinTrap™
Affinity chromatography columns, Ab SpinTrap™
Ab SpinTrap™, 34 µm, 100 µl
Price
751,00 €
Log in voor beschikbaarheid
1 * 50 ST
Product Details & Documents
Ab SpinTrap columns are prepacked, single-use spin columns for rapid purification of monoclonal and polyclonal antibodies from serum and cell culture supernatants. Designed for small-scale purification of multiple samples in parallel.
  • Rapid and convenient small-scale purification of IgG
  • Protein G Sepharose High Performance in prepacked spin columns
  • No filtration or centrifugation of different crude samples necessary for antibody purification
  • Classic and crosslink protocols for protein enrichment (immunoprecipitation)
  • Elution conditions formatted for both LC-MS and electrophoresis analysis
The spin columns are also suitable for use in antibody screening experiments. Ab SpinTrap columns are used together with a standard microcentrifuge and one purification run is performed in less than 20 min. Ab SpinTrap contains Protein G Sepharose High Performance, which has high protein binding capacity and is compatible with all commonly buffers used in antibody purification with immobilized Protein G media. For optimal performance in antibody purification, use Ab SpinTrap together with buffers prepared using Ab Buffer Kit.
Designed for small-scale purification of multiple samples in parallel. Ab SpinTrap is designed for efficient, small-scale purification of monoclonal and polyclonal antibodies from serum and cell culture supernatants. In addition, a protocol for immunoprecipitation is included.
Specificaties
  • Partikelgrootte:
    34 µm
  • Capaciteit:
    100 µl
  • Omschrijving:
    Ab SpinTrap™
  • Seq:
    01
  • Cat. No.:
    28-4083-47
  • Bindingscapaciteit:
    > 1 mg IgG/column