Bottles, wide neck, PE, rectangular, with screw cap
1000 ml, 58 mm, 103×80×176 mm
Catalogus # SEMA0063
Leverancier: SEMADENI
Ga naar:
Product Details & Documents
HDPE, translucent.
- Good chemical resistance
- Good temperature resistance
Specificaties
- Capaciteit:1000 ml
- b×d×h:103×80×176 mm
- Ø inw. hals:58 mm