ortho-Fosforzuur 0.1 mol/l (0.3 N)
Catalog # PRLS88168.9010
Supplier: PROLABO CMP
CAS Number:
Specifications
- Verpakking:Plastic container
- VPE:10 L
- Seq:5214
Specifications
About this item
:
:98 g/mol
Storage Temperature:Kamertemperatuur
EINECS:231-633-2
UN:1805
ADR:8,III
CAS Number:7664-38-2
Formula:H₃PO₄
MDL Number:MFCD00011340