Laboratory jacks
Leverancier: R L ENTERPRISES
Ga naar:
Over dit artikel
These lab jacks are manufactured to close tolerances from aluminium with stainless steel operating screw.
- Top and bottom plates are specially designed aluminium extrusion giving exceptional rigid and robust support without excessive weight
- Plates are blue anodised finish
- Designed to be completely stable and free from rocking movements
6 Opties Hieronder Beschikbaar
Product Details & Documents
These lab jacks are manufactured to close tolerances from aluminium with stainless steel operating screw.
- Top and bottom plates are specially designed aluminium extrusion giving exceptional rigid and robust support without excessive weight
- Plates are blue anodised finish
- Designed to be completely stable and free from rocking movements
Product Family Options
Productinformatie
- PlaatgrootteMax. statische belastingWerkhoogteKleurBeschikbaarheidPrice