Window thermometer
−50…+50 °C, 200 mm, Blue spirit
Catalogus # 620-1181
Leverancier: GLASWARENFABRIK KARL HECHT
Ga naar:
Product Details & Documents
Vienna type window thermometer with metal fittings.
Specificaties
- Lengte:200 mm
- Vulling:Blue spirit
- Meetbereik:−50…+50 °C