Wall-mounting room thermometer
−30…+50 °C, 200 mm, Coloured filling
Catalog # HECH43513010
Supplier: GLASWARENFABRIK KARL HECHT
Jump to:
Product Details & Documents
This room thermometer is made of beech wood with natural varnish.
Services: Voor installatie, IQOQ, erkende kalibratie onder ISO17025, onderhoud en reparatie mail naar [email protected]
Specifications
- Lengte:200 mm
- Vulling:Coloured filling
- Meetbereik:−30…+50 °C