Orders LinkContactUsLinkComponent
Workbenches, Modulab
Plain dry bench, 600 mm base frame, with Trespa Athlon on aircraft plywood coating, 600×600×900 mm
Catalog # 132-7745
Supplier:   EQUIP LABO
Workbenches, Modulab
Catalog # 132-7745
Supplier:  EQUIP LABO
Workbenches, Modulab
Workbenches, Modulab
Plain dry bench, 600 mm base frame, with Trespa Athlon on aircraft plywood coating, 600×600×900 mm
Price
486,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Frame:
30×30-mm metal base frame, protected by a white baked epoxy resin, fitted with level adjustment gas lift columns
Rear strip and a fluid back panel, with water resistant melamine on both sides, with all borders veneered
Metallic tube stop bar protected by a white epoxy resin
  • 30 x 30-mm metal base frame, protected by a white baked epoxy resin, fitted with level adjustment gas lift columns
  • Rear strip and a fluid back panel, with water resistant melamine on both sides, with all borders veneered
  • Metallic tube stop bar protected by a white epoxy resin
Front strip:
Laminate Oslo blue (on request: white, yellow, broom or grey)
Worktop with five possible surfaces:
White 12/10 post-formed laminate on the front edge
Trespa® Athlon® resin (thickness: 8 mm) on chipboard support (thickness: 22 mm), white surface and a PVC border
Stoneware slab (thickness: 8 mm) on a water-resistant support (thickness: 22 mm) with a PVC border
Tempered unleaded enamelled glass — 'ice' colour — with a thickness of 6 mm on a water resistant chipboard support (thickness: 22 mm), a white PVC border (thickness: 4 mm), flexible seal between the glass and the support
Bare benches (composition):
1 fluid back panel
2 metal legs
1 dry worktop
1 laminate front strip
1 rear strip
1 stop bar
2 or 3 extensions for 750 mm deep benches with 600 mm legs
The 750 mm deep bench is equipped as standard with a 600 mm base, which cannot accommodate a splashback shelf. To add a splashback shelf, a 750 mm base is required.
Dry benches with splashback shelf (composition):
1 fluid back panel
2 metal legs
1 dry worktop
1 laminate front strip
1 rear strip
1 splashback, water resistant melamine on both sides, on a high density chipboard support, which can accommodate plumbing fixtures and electrical sockets
2 or 3 veneer shelf supports
1 white, water resistant melamine shelf on two sides, Oslo blue borders, 160 mm width (10 mm front edge)
1 metallic stop bar
Conforms to the AFNOR 15201 standard.
 : Conforms to the AFNOR 15201 standard.
 : Supplied as a ready-to-assemble, flat-pack kit. The mounting fittings as well as the delivery fittings are automatically provided for all standard benches.
Specifications
  • Omschrijving:
    Plain dry bench, 600 mm base frame, with Trespa Athlon on aircraft plywood coating
  • Seq:
    004
  • b×d×h:
    600×600×900 mm