Orders LinkContactUsLinkComponent
Entomological forceps
Entomological forceps
Gebogen, 120 mm
Catalogus # 232-0038
Leverancier:   MURRAY S
Entomological forceps
Catalogus # 232-0038
Leverancier:  MURRAY S
Entomological forceps
Entomological forceps
Gebogen, 120 mm
Price
11,50 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Stainless steel. Forceps for the collection of entomological specimens.
  • Guide pin helps to keep the tips aligned
  • Wide tips help to avoid damages of the specimen
  • Curved tips won't crush delicate larvae or eggs
Specificaties
  • Type:
    Gebogen
  • Lengte:
    120 mm