Counting chambers, Jessen
HECH40450702
- Kamerdiepte:0,4 mm
- Type:Jessen
- Omschrijving:Counting chamber (CE)
- Seq:001
- Environmentally Preferable:
- VPE:1
These double ruling chambers are used for blood tests, urine analysis, and counting cells in cerebrospinal fluid (CSF).