Spuitflessen, smalle hals
KAUT76083
: Kautex
- VPE:1
LDPE, translucent with LDPE spray cap.
- Good chemical resistance
- Heat resistant up to 80 °C
- Leakproof
Economical, soft and very easy to squeeze. For use with general solutions such as water, soap, dilute acids and dilute alkalis. The tip can be cut off to increase the flow rate.