Cell culture plates with Cell+ surface
Catalogus # SARS83.1836.300
Leverancier: SARSTEDT
Specificaties
- Aant. wells:24
- Verpakt:1 per zak
- Omschrijving:Cell culture plates, Cell+ growth surface
- Cultuuroppervlak:2 cm²
- VPE:50
- Wellvolume:3,6 ml
Specificaties
Over dit artikel
PS, gamma sterilised, with lid.
- Wells with raised rims
- Stackable
- Lid with condensation rings
- Lid can only be positioned in one direction, avoids cross contamination
Cell+ TC plates for the growth of primary cultures and other fastidious cell lines.