Orders LinkContactUsLinkComponent
Quantus AreaDress | Avantor
Quantus AreaDress
basan_Quan_Areadress_200.jpg
Features top:
  • Two-way front zipper (covered)
  • High collar with press stud
  • Comfortable Raglan style
  • Antistatic knitted cuffs
  • Stitched collar patch with label
  • Hanger in collar
Colour: White, further colours upon request
Fabric: 100% Polyester with Carbon
Sizes: XS - XXXL
Features trousers:
  • Wide, double stitched comfort waist band
  • Knitted cuffs at ankle height
Colour: White, further colours upon request
Fabric: 100% Polyester with Carbon
Sizes: XS - XXXL