ECHO CellCyte 1 Live Cell Imaging and Analysis
ECHO CellCyte 1 Live Cell Imaging and Analysis
CELLCYTE 1 complete imaging system
N° de catalogue ECHLECCC1200
New Product
Fournisseur:   DISCOVER ECHO
ECHO CellCyte 1 Live Cell Imaging and Analysis
N° de catalogue ECHLECCC1200
Fournisseur:  DISCOVER ECHO
New Product
ECHO CellCyte 1 Live Cell Imaging and Analysis
CELLCYTE 1 complete imaging system
Prix
68 710,00 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Live cell imaging and analysis in real-time from within the incubator.
  • Enhanced contour: contrast rich transmission imaging mode
    CellCyte™ studio: automated acquisition and user friendly image and data visualization software
    CellCyte™ analysis: integrated real-time image analysis using smart algorithms
    Open design: quick access to cells for experimentation and easier maintenance
    Export formats: Movie (AVI), Image (TIFF, PNG), Data (CSV, XLXS)
5 MP monochrome camera - increases field of view and resolution for fluorescence imaging. Operating conditions - (Imaging Unit) 10 to 40 °C, RH up to 95%, (Controller Unit) 20 to 40 °C. Spheroid Software (optional) - real time spheroid imaging and analysis at 4x.
Illumination technique: Enhanced contour and Fluorescence imaging (three Fluorescence channels included). 10TB internal storage (Controller Unit). Additional storage can be added via external hard drives, allowing users to transfer information between control units. Ultra-bright LEDs with 50000 - hour lifespan. Intuitive software drive the CELLCYTE™ Controller and Imaging units, allowing users to image and analyze experiments across multiple devices.
The Cellcyte 1 is suitable for a variety of cell culture vessels and can support up to 6 wellplates (6 to 384 wells each) and T-series flasks.
Ordering information: To be completed with Dell Ultrasharp 27" 4K display, HP Z2 GD Mini PC, and flask inserts, which are sold separately.
Delivery information: Supplied with CellCyte 1 imaging unit, controller unit, integrated monochrome camera for enhanced contour and fluorescence imaging, Olympus® 10× objective, and power supply.
Spécifications
  • Désignation:
    CELLCYTE 1 complete imaging system