Acide 3,4-dihydroxybenzoïque
Bottle
Catalog # APOSOR52125-2.5KG
New Product
Supplier: Apollo Scientific
Acide 3,4-dihydroxybenzoïque
Catalog # APOSOR52125-2.5KG
Supplier: Apollo Scientific
New Product
CAS Number: 99-50-3
Jump to:
Product Details & Documents
Synonyms: Protocatechuic acid
- CAS Number:99-50-3
- Formula:C₇H₆O₄
- MDL Number:MFCD00002509
- Molecular Weight:154.12 g/mol
- Melting Pt:197…200 °C (déc.)
- Storage Temperature:Température ambiante
- EINECS:202-760-0
Specifications
Specifications
- Type de conditionnement:Bottle



