ContactUsLinkComponent
,,14 components: 100ug/ml each of Al As Ca Cd Cr Cu Fe Hg K Mg Na Ni Pb Zn in HNO3 2% ,,CRM, ISO 17034 and ISO 17025,,,, ,,Conservation 24 months.,,,, ,,Store under normal laboratory conditions, at temperatures between 15C 1 * 1 L
N° de catalogue CMPLRM1105921L
New Product
,,14 components: 100ug/ml each of Al As Ca Cd Cr Cu Fe Hg K Mg Na Ni Pb Zn in HNO3 2% ,,CRM, ISO 17034 and ISO 17025,,,, ,,Conservation 24 months.,,,, ,,Store under normal laboratory conditions, at temperatures between 15C 1 * 1 L
N° de catalogue CMPLRM1105921L