Human Leukopak
Human Leukopak
Human leukopak, healthy donor, 5B cells (5×10⁹ cells/bag), Cryopreserved, Single donor, 1
N° de catalogue PRTV12192100050
Fournisseur:   PROVITRO
Human Leukopak
N° de catalogue PRTV12192100050
Fournisseur:  PROVITRO
Numéro CAS:  
Human Leukopak
Human Leukopak
Human leukopak, healthy donor, 5B cells (5×10⁹ cells/bag), Cryopreserved, Single donor, 1
Numéro CAS:  
Prix
4 360,00 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Our highly cell-concentrated Leukopak is a specialised product from leukapheresis using the Spectra Optia system. This advanced technology employs centrifugal mechanisms for continuous blood flow, extracting white blood cells from peripheral blood into a sterile bag with ACD-A anticoagulant. The enriched product includes monocytes, lymphocytes, platelets, plasma, and erythrocytes.
  • Leukopak is collected from healthy donors using approved permission forms and procedures
  • Collection procedures have been approved by the Institutional Review Board (IRB)
  • The product includes donor information and serology testing for: HIV, HBs-AG, HCV
  • Additional optional information and testing are available for a small fee
Our offerings encompass Leukopaks sourced from our existing inventory, alongside the provision for prospective collections tailored to specific research inquiries. This versatile range ensures a breadth of options to suit a variety of research requirements, whether drawing from our readily available stock or arranging for custom collections based on unique project needs.
Storage: Prepare cells for extended storage in the vapor phase of liquid nitrogen or thaw them for immediate use. Avoid storing cells in the liquid phase of nitrogen. Short-term storage (less than two weeks) at −80 °C is acceptable but should be minimised for optimal stability. Once thawed, it's crucial to use the samples immediately.
Attention: Not intended for any human or animal diagnostic or therapeutic use.
Spécifications
Spécifications
  • Format de cellule:
    Cryopreserved
  • Cdt:
    1
  • Donneur unique/donneurs regroupés:
    Single donor
  • Désignation:
    Human leukopak, healthy donor, 5B cells (5×10⁹ cells/bag)
Spécifications