Antibiotic Sensitivity Discs
Cefoxitin, 30 mcg, 50
N° de catalogue HIMESD041-5X50DS
Fournisseur:   HIMEDIA
Antibiotic Sensitivity Discs
N° de catalogue HIMESD041-5X50DS
Fournisseur:  HIMEDIA
Numéro CAS:  
Antibiotic Sensitivity Discs
Cefoxitin, 30 mcg, 50
Numéro CAS:  
Prix
19,50 €
Connexion Pour la disponibilité
1 * 50 ST
Product Details & Documents
Antibiotic sensitivity disc are used to determine the effectivness of an antibiotic against a microorganism as a guide to antibiotic therapy.
The disc consist of a specific concentration of an antibiotic in mcg which is tested against a microorganism of a standard density, swabbed uniformly on an appropriate media following overnight incubation.
The antibiotic is impregnated on a filter paper disc in specified concentration. Both sides of the disc are printed with name and concentration for easy identification.
Spécifications
Spécifications
  • Cdt:
    50
  • Désignation:
    Cefoxitin, 30 mcg
Spécifications