ContactUsLinkComponent
,,25 components: P 250mg/l Al 100mg/l Ba 100mg/l B 50mg/l Fe 50mg/l Zn 50mg/l Mn 10mg/l Mo 10mg/l Co 5mg/l Cr 5mg/l Cu 5mg/l Ni 5mg/l Pb 5mg/l Sb 5mg/l Sn 5mg/l Te 5mg/l Ti 5mg/l V 5mg/l Ag 2.5mg/l As 2.5mg/l S 1 * 100 mL
Catalog # CMPLRM112228L1
New Product
,,25 components: P 250mg/l Al 100mg/l Ba 100mg/l B 50mg/l Fe 50mg/l Zn 50mg/l Mn 10mg/l Mo 10mg/l Co 5mg/l Cr 5mg/l Cu 5mg/l Ni 5mg/l Pb 5mg/l Sb 5mg/l Sn 5mg/l Te 5mg/l Ti 5mg/l V 5mg/l Ag 2.5mg/l As 2.5mg/l S 1 * 100 mL
Catalog # CMPLRM112228L1