ReadiPrep™ Exosome Isolation Kit
200 preparations, 200
N° de catalogue AATB60205
Fournisseur:   AAT Bioquest
ReadiPrep™ Exosome Isolation Kit
N° de catalogue AATB60205
Fournisseur:  AAT Bioquest
Numéro CAS:  
ReadiPrep™ Exosome Isolation Kit
200 preparations, 200
Numéro CAS:  
Prix
620,00 €
Connexion Pour la disponibilité
1 * 200 ST
Product Details & Documents
Designed for consistent and dependable results. The ReadiPrep™ exosome isolation kit offers a streamlined and effective approach to isolating intact exosomes from diverse biological samples, including cell culture media and serum.
  • Robust protocol
  • Produced exosomes are suitable for further downstream analyses or purification via affinity-based methods
  • Six-month shelf life
The principle behind this kit is based on the selective precipitation of exosomes by inducing the removal of water molecules from the solution.
 : Shipped at RT.
 : Research use only.
Spécifications
Spécifications
  • Cdt:
    200
  • Info cdt:
    200 preparations
Spécifications