ReadiPrep™ Exosome Isolation Kit
50 preparations, 50
Catalogus # AATB60204
Leverancier:   AAT Bioquest
ReadiPrep™ Exosome Isolation Kit
Catalogus # AATB60204
Leverancier:  AAT Bioquest
CAS-nummer:  
ReadiPrep™ Exosome Isolation Kit
50 preparations, 50
CAS-nummer:  
Price
268,00 €
Log in voor beschikbaarheid
1 * 50 ST
Product Details & Documents
Designed for consistent and dependable results. The ReadiPrep™ exosome isolation kit offers a streamlined and effective approach to isolating intact exosomes from diverse biological samples, including cell culture media and serum.
  • Robust protocol
  • Produced exosomes are suitable for further downstream analyses or purification via affinity-based methods
  • Six-month shelf life
The principle behind this kit is based on the selective precipitation of exosomes by inducing the removal of water molecules from the solution.
 : Shipped at RT.
 : Research use only.
Specificaties
Specificaties
  • Cdt:
    50
  • Info cdt:
    50 preparations
Specificaties