Erlenmeyer Flasks with screw cap
250 ml, GL 45
N° de catalogue NEUB0748-10-025045
Fournisseur: NEUBERT VOLUME GLASSWAERE
Erlenmeyer Flasks with screw cap
N° de catalogue NEUB0748-10-025045
Fournisseur: NEUBERT VOLUME GLASSWAERE
Aller à :
Product Details & Documents
Borosilicate glass 3.3, clear.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Volume graduations and labelling area. Screw cap with PTFE coated seal.
Spécifications
- Filetage:GL 45
- Capacité:250 ml
- Seq:01
- Cat. No.:NEUB0748-10-025045