Storage Cleaners Cabinets
Cleaners cabinet on casters, 2, Acier inoxydable 304, 500×800×1800 mm
N° de catalogue PALBJC0850C
Fournisseur: Palbam
Aller à :
Product Details & Documents
Designed for storing mops and cleaning materials.
- Two doors
Fabrication material stainless steel 304 with electropolished finish.
Spécifications
- Désignation:Cleaners cabinet on casters
- Matière:Acier inoxydable 304
- Seq:01
- l×p×h:500×800×1800 mm
- Portes:2