Nullscript ≥97% (par CCM)
1 mg
N° de catalogue ENZOBMLGR3270001
Fournisseur: Enzo Life Sciences
Nullscript ≥97% (par CCM)
N° de catalogue ENZOBMLGR3270001
Fournisseur: Enzo Life Sciences
Numéro CAS: 300816-11-9
Aller à :
Product Details & Documents
HDAC inhibitor
Negative control compound for the histone deacetylase (HDAC) inhibitor scriptaid (Prod. No. BML-GR326).
Synonymes: 4-(1,3-Dioxo-1H,3H-benzo[de]isoquinolin-2-yl)-N-hydroxybutanamide
- Numéro CAS:300816-11-9
- Formule:C₁₆H₁₄N₂O₄
- Numéro MDL:MFCD00405081
- Poids moléculaire:298.30 g/mol
Spécifications
Spécifications
- Cdt:1 mg
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



