Powder spatulas
Powder spatulas
Flexible, double-ended, one standard spatula end, PTFE coated, 150 mm
Catalog # HAMMHSN075-15T
Supplier:   HAMMACHER KARL
Powder spatulas
Catalog # HAMMHSN075-15T
Supplier:  HAMMACHER KARL
Powder spatulas
Powder spatulas
Flexible, double-ended, one standard spatula end, PTFE coated, 150 mm
Price
28,20 €
Log in for availability
1 * 1 ST
Product Details & Documents
Flexible, double-ended spatulas for powder handling.
Specifications
  • Désignation:
    Flexible, double-ended, one standard spatula end, PTFE coated
  • Longueur:
    150 mm
  • Seq:
    02