Biotin-dT Phosphoramidite
Product Details & Documents
Biotin-dT phosphoramidite provides a valuable tool to add the biotin tag to a desired location of an oligo sequence.
- DMT group allows coupling quantification and can be used in RP cartridge and HPLC purification techniques
- Thoroughly referenced
Biotin-dT phosphoramidite contains a DMT group to allow quantification of coupling. It can be inserted into a target sequence as a replacement for a dT residue.
It is complimentary to the commonly used biotin phosphoramidite that contains no DMT group and can only be added at the 5'-terminus, thus terminating synthesis.
Emballage: Plastic vial.
Attention: For research use only.
- Numéro CAS:198080-40-9
- Formule:C₆₉H₈₉N₈O₁₂PS
- Poids moléculaire:1285.55 g/mol
Spécifications
Spécifications
- Cdt:50 µmol
Spécifications
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



