PD-10 Buffer Reservoir
PD-10 Buffer Reservoir
PD-10 buffer reservoir
N° de catalogue AATB60508
Fournisseur:   AAT Bioquest
PD-10 Buffer Reservoir
N° de catalogue AATB60508
Fournisseur:  AAT Bioquest
PD-10 Buffer Reservoir
PD-10 Buffer Reservoir
PD-10 buffer reservoir
Prix
177,00 €
Connexion Pour la disponibilité
1 * 5 ST
Product Details & Documents
PD-10 Buffer Reservoir provides an easy and rapid way for equilibrating PD-10 columns.
  • Efficient and convenient
  • Simple to use
  • Improves equilibration and elution procedures
Attention: For research use only.
Spécifications
  • Désignation:
    PD-10 buffer reservoir
  • Seq:
    01
  • Cat. No.:
    AATB60508