Wire Baskets for Test Tubes
120×120×120 mm
N° de catalogue LINK51108211212
Fournisseur: LINKER INDUSTRIE TECHN
Aller à :
Product Details & Documents
Wire basket of welded mesh w 8 mm, d 1 mm for test tubes and more. Made up of stainless steel W-no. 1.4301, cube shape, electrochemically polished.
Spécifications
- Seq:02
- l×p×h:120×120×120 mm