Acétone ≥99.5%, GPR RECTAPUR®
950 L
Fournisseur: VWR Chemicals
Acétone ≥99.5%, GPR RECTAPUR®
N° de catalogue 20065.950
Fournisseur: VWR Chemicals
Numéro CAS: 67-64-1
Aller à :
Product Details & Documents
Synonymes: 2-Propanone, DMK, Propanone, Dimethyl ketone
- Numéro CAS:67-64-1
- Formule:CH₃COCH₃
- Numéro MDL:MFCD00008765
- Poids moléculaire:58.08 g/mol
- Index Merck:13,00067
- Point d'ébullition:56,2 °C (1013 hPa)
- Point de fusion:–95,4 °C
- Densité:0,792 g/cm³ (20 °C)
- Point d'éclair:<–20 °C (coupe fermée)
- Température de stockage:Température ambiante
- EINECS:200-662-2
- UN:1090
- ADR:3,II
- REACH:01-2119471330-49
Documents
Certificats
Rechercher par numéro de lot1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000) 2,5L Plastic bottle for liquids DIN 45 UN X (ZPACK13962500) 225L Metal drum UN (ZPACE8010050) 25L Plastic drum for liquids DIN 61 (ZPACK13952500) 30L Metal drum for liquids UN X (ZPACK12124125)
Packaging Information
Plus
20065_ACETONE GPR RECTAPUR_BSE_REV1
BSE/TSE Certificates
P20_VWRC20065.293_SPEC_FR.pdf P20_VWRC20065.327_SPEC_FR.pdf P20_VWRC20065.362_SPEC_FR.pdf P20_VWRC20065.464_SPEC_FR.pdf P20_VWRC20065.470_SPEC_FR.pdf
Product specification
Plus
VWR Chemicals Big Packs and Bulks
Product Literature
Spécifications
Résultats des tests de spécification
- Related substances:Passes test
- Boiling point:56 - 56.7 °C
- Density (20/4):0.79 - 0.793
- Evaporation residue:Max. 20 ppm
- Assay:Min. 99.5 %
- Acidity:Passes test
- Alkalinity:Passes test
- Appearance of solution:Passes test
- Insolubility in water:Passes test
- IR Spectrum:Passes test
- Substances reducing KMnO4 (as O):Max. 2 ppm
- Water:Max. 0.3 %
Spécifications
- Cdt:950 L
- Cat. No.:20065.950
- Certification environnementale:Chem21 Solvent Selection Guide recommended after discussion
- Environmentally Preferable:Safer Human and Environmental Health
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



