Code Corporation CR2700 Barcode Scanner Accessories
Code Corporation CR2700 Barcode Scanner Accessories
Rechargeable lithium-ion battery, dark grey, 12,7×63,5×76,2 mm, for model CR2700
N° de catalogue BRDY176699
Fournisseur:   Brady
Code Corporation CR2700 Barcode Scanner Accessories
N° de catalogue BRDY176699
Fournisseur:  Brady
Code Corporation CR2700 Barcode Scanner Accessories
Code Corporation CR2700 Barcode Scanner Accessories
Rechargeable lithium-ion battery, dark grey, 12,7×63,5×76,2 mm, for model CR2700
Prix
98,80 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Accessories available for the Barcode Scanner CR2700.
Cable (BRDY176508) - Use this 6-foot straight cable for making USB to Micro USB connections. Reader termination: Micro USB. Host termination: USB Type A.
B27 lithium-ion battery (BRDY176699) - powers the CR2700 reader with a unique set of features that keep workflow interruptions to a minimum. Power gauge to track battery level. Quick-release latch allows for toolless swapping. The B27 battery can charge while installed in a CR2700 reader using the inductive charging station (CRA-A271).
Attention: Model BRDY176699 has lithium-ion battery - must ship ground.
Spécifications
  • Désignation:
    Rechargeable lithium-ion battery, dark grey, 12,7×63,5×76,2 mm, for model CR2700
  • Seq:
    02