Measuring Cables, CYK10
Measuring cable CYK10, wire terminals; Nonhazardous area approval, 10 m
N° de catalogue ENDRCYK10-A101
Fournisseur: Endress+Hauser
Aller à :
Product Details & Documents
CYK10 measuring cable is used with all sensors with Memosens plug-in head. Forget about moisture and benefit from completely safe signal transmission.
- Memosens data cable for all sensors with Memosens plug-in head
- Maximum process safety via non-contact, inductive and digital signal transmission
- No contact corrosion, absolutely watertight, no interference potential
- Increased availability of the measuring point: Digital data transmission provides an automatic error message if the signal is lost
CYK10 guarantees absolute water-proof connection of Memosens sensors to the transmitter. Its inductive coupling shows no contact corrosion and guarantees galvanic isolation of transmitter and medium. Simply forget about limited cable length, interfering potentials or moisture and benefit from completely safe and reliable signal transmission.
Documents
TDS Measuring Cables CYK10
Fiche technique
Spécifications
- Désignation:Measuring cable CYK10, wire terminals; Nonhazardous area approval
- Longueur:10 m
- Seq:03