Guard Column Holders
Guard Column Holders
UHPLC/HPLC guard column holder for direct coupling of 5 mm guard cartridges (zero dead volume) to analytical columns, suitable for XELA and XCORE cartridges, can be mounted on analytical columns of most manufacturers
N° de catalogue ISERAA13-000007
Fournisseur:   ISERA
Guard Column Holders
N° de catalogue ISERAA13-000007
Fournisseur:  ISERA
Guard Column Holders
UHPLC/HPLC guard column holder for direct coupling of 5 mm guard cartridges (zero dead volume) to analytical columns, suitable for XELA and XCORE cartridges, can be mounted on analytical columns of most manufacturers
Prix
154,00 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Spécifications
  • Désignation:
    UHPLC/HPLC guard column holder for direct coupling of 5 mm guard cartridges (zero dead volume) to analytical columns, suitable for XELA and XCORE cartridges, can be mounted on analytical columns of most manufacturers
  • Seq:
    07