Guard Column Holders
UHPLC/HPLC guard column holder for direct coupling of 5 mm guard cartridges (zero dead volume) to analytical columns, suitable for XELA and XCORE cartridges, can be mounted on analytical columns of most manufacturers
N° de catalogue ISERAA13-000007
Fournisseur: ISERA
Aller à :
Product Details & Documents
Spécifications
- Désignation:UHPLC/HPLC guard column holder for direct coupling of 5 mm guard cartridges (zero dead volume) to analytical columns, suitable for XELA and XCORE cartridges, can be mounted on analytical columns of most manufacturers
- Seq:07