Eau, GPR RECTAPUR®
200 L
Fournisseur: VWR Chemicals
Aller à :
Product Details & Documents
Synonymes: Hydrogen oxide
- Numéro CAS:7732-18-5
- Formule:H₂O
- Numéro MDL:MFCD00011332
- Poids moléculaire:18.02 g/mol
- Index Merck:13,10098
- Point d'ébullition:100 °C (1013 hPa)
- Point de fusion:0 °C
- Densité:1 g/cm³ (4 °C)
- Température de stockage:Température ambiante
- EINECS:231-791-2
Documents
Certificats
Rechercher par numéro de lot10L Cubitainer UN (ZPACK13260004) 10L Plastic drum for liquids DIN 51 UN (ZPACK13951000) 25L Plastic drum for liquids DIN 61 (ZPACK13952500)
Packaging Information
P20_VWRC83612.360_SPEC_FR.pdf P20_VWRC83612.400_SPEC_FR.pdf P20_VWRC83612.410_SPEC_FR.pdf P20_VWRC83612.460_SPEC_FR.pdf P20_VWRC83612.550_SPEC_FR.pdf
Product specification
Plus
VWR Chemicals Big Packs and Bulks
Product Literature
Spécifications
Résultats des tests de spécification
- Acidity or alkalinity:Passes test
- Oxidisable substances:Passes test
- Conductivity (20°C):Max. 1.5 µS/cm
- Evaporation residue:Max. 1 ppm
- Heavy metals (as Pb):Max. 0.1 ppm
- Cl (Chloride):Max. 0.1 ppm
- NH4 (Ammonium):Max. 0.2 ppm
- NO3 (Nitrate):Max. 0.2 ppm
- SO4 (Sulphate):Max. 1 ppm
- Ca (Calcium):Max. 0.1 ppm
- Na (Sodium):Max. 0.1 ppm
Spécifications
- Cdt:200 L
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



