Plum iBox
Plum iBox
Plum iBox 2 with 1×200 ml Plum Eyewash pH Neutral 4,9% phosphate and 1×500 ml Plum Eyewash 0.9% sodium chloride
N° de catalogue PLUM4921
Fournisseur:   Plum Safety
Plum iBox
N° de catalogue PLUM4921
Fournisseur:  Plum Safety
Plum iBox
Plum iBox
Plum iBox 2 with 1×200 ml Plum Eyewash pH Neutral 4,9% phosphate and 1×500 ml Plum Eyewash 0.9% sodium chloride
Prix
66,90 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
The new Plum iBox is balancing key features in a modern design and is built in a strong and durable material. See-through window lets users keep track of expiry dates immediately.
  • Clearly visible eyewash bottles
  • Quick and efficient overview of expiry dates and product condition
  • Grab from the wall bracket and quickly bring to injured person
  • Spacious design allows access to bottles fast and easily
  • Easily opens – mounted on the wall or standing on the floor
  • Strong and durable material (EPP)
Release Plum iBox from the wall and quickly bring the eyewash to the injured person. Delivered with wall bracket, eyewash plan, and wall mounting set.
Plum iBox 1 iss uitable for dusty environments and small workplaces, with a risk of getting foreign bodies into the eyes. Plum iBox 2 is suitable for workplaces where there is a risk of getting both foreign bodies and/or acids and alkali in the eye. Plum iBox 3 a comprehensive solution for eyewash. Particularly suitable for workplaces where there is a risk of getting both foreign bodies and acids and alkali in the eye.
Certifications: Install the eyewash station less than 20 m or less than 10 seconds from chemical hazard according to EN 15154-4 recommendation.
Spécifications
  • Désignation:
    Plum iBox 2 with 1×200 ml Plum Eyewash pH Neutral 4,9% phosphate and 1×500 ml Plum Eyewash 0.9% sodium chloride
  • Seq:
    03
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.