ContactUsLinkComponent
Specifications:- Capillary Column Phase: DB-WAX Ultra Inert. Capillary Tubing: Fused Silica. Film Thickness: 0.5 um. GC Column Forma t: Intuvo. GC Column Type: Ultra Inert. Inner Diameter (ID): 0.32 mm. Length: 60 m. Polarity: High Polarity. With Sma 1 * 1 Pce
Catalog # HEWL123-7063UI-INT
Supplier:  Agilent
Specifications:- Capillary Column Phase: DB-WAX Ultra Inert. Capillary Tubing: Fused Silica. Film Thickness: 0.5 um. GC Column Forma t: Intuvo. GC Column Type: Ultra Inert. Inner Diameter (ID): 0.32 mm. Length: 60 m. Polarity: High Polarity. With Sma 1 * 1 Pce
Catalog # HEWL123-7063UI-INT
Supplier:  Agilent
Frequently Bought Together