ContactUsLinkComponent
J&W DB-5ms Ultra Inert Intuvo GC column module, 60 m, 0.25 mm, 0.25 um. (compatible only with the Intuvo 9000 GC System). Specificat ions:- Capillary Column Phase: DB-5ms Ultra Inert. Capillary Tubing: Fused Silica. Film Thickness: 0.25 um. GC Column 1 * 1 Pce
N° de catalogue HEWL122-5562UI-INT
Fournisseur:  Agilent
undefined
J&W DB-5ms Ultra Inert Intuvo GC column module, 60 m, 0.25 mm, 0.25 um. (compatible only with the Intuvo 9000 GC System). Specificat ions:- Capillary Column Phase: DB-5ms Ultra Inert. Capillary Tubing: Fused Silica. Film Thickness: 0.25 um. GC Column 1 * 1 Pce
N° de catalogue HEWL122-5562UI-INT
Fournisseur:  Agilent
Frequently Bought Together