ContactUsLinkComponent
Specifications:- Capillary Column Phase: DB-35ms Ultra Inert. Capillary Tubing: Fused Silica. Film Thickness: 0.25 um. GC Column For mat: Intuvo. GC Column Type: Ultra Inert. Inner Diameter (ID): 0.25 mm. Length: 30 m. Polarity: Mid Polarity. With Sm 1 * 1 Pce
N° de catalogue HEWL122-3832UI-INT
Fournisseur:  Agilent
undefined
Specifications:- Capillary Column Phase: DB-35ms Ultra Inert. Capillary Tubing: Fused Silica. Film Thickness: 0.25 um. GC Column For mat: Intuvo. GC Column Type: Ultra Inert. Inner Diameter (ID): 0.25 mm. Length: 30 m. Polarity: Mid Polarity. With Sm 1 * 1 Pce
N° de catalogue HEWL122-3832UI-INT
Fournisseur:  Agilent
Frequently Bought Together