Agilent J&W DB-Select 624 Ultra Inert Intuvo GC column module, 30 m, 0.25 mm, 1.40 um. Specifications:- Capillary Column Phase: DB-S elect 624 UI for 467 . Capillary Tubing: Fused Silica. Film Thickness: 1.4 um. GC Column Format: Intuvo. GC Column Ty 1 * 1 Pce
Catalog # HEWL122-0334UI-INT
Supplier: Agilent
Frequently Bought Together