Specifications:- Capillary Column Phase: HP-5ms Ultra Inert. Capillary Tubing: Fused Silica. Film Thickness: 0.25 um. GC Column Form at: 7 inch. GC Column Type: Ultra Inert. Inner Diameter (ID): 0.25 mm. Length: 30 m. Polarity: Low Polarity. With Sma 1 * 1 Pce
N° de catalogue HEWL19091S-433UI-K
Fournisseur: Agilent
Frequently Bought Together