InfinityLab Poroshell 120 EC-CN, 4.6 x 50 mm, 2.7 um, analytical LC column, with column ID. Specifications:- Carbon Load: 3.5 %. End capped: Double. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 4.6 mm. LC Column Brand: InfinityLab P 1 * 1 Pce
N° de catalogue HEWL699975-905T
Fournisseur: Agilent
Frequently Bought Together