InfinityLab Poroshell 120 HILIC, 4.6 x 50 mm, 2.7 um, analytical LC column, with column ID. Specifications:- Carbon Load: 0 %. Endca pped: No. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 4.6 mm. LC Column Brand: InfinityLab Poroshe 1 * 1 Pce
Catalog # HEWL699975-901T
Supplier: Agilent
Frequently Bought Together