InfinityLab Poroshell HPH-C8, 4.6 x 50 mm, 2.7 um, analytical LC column, with column ID. Specifications:- Endcapped: Yes. Guard Colu mn: No. Hardware: Conventional. Inner Diameter (ID): 4.6 mm. LC Column Brand: InfinityLab Poroshell 120. LC Column Ma 1 * 1 Pce
N° de catalogue HEWL699975-706T
Fournisseur: Agilent
Frequently Bought Together