ContactUsLinkComponent
InfinityLab Poroshell HPH-C18, 2.1 , 50 mm, 2.7 um, narrow bore LC column, with column ID. Specifications:- Endcapped: Yes. Guard Co lumn: No. Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab Poroshell 120. LC Column 1 * 1 Pce
N° de catalogue HEWL699775-702T
Fournisseur:  Agilent
InfinityLab Poroshell HPH-C18, 2.1 , 50 mm, 2.7 um, narrow bore LC column, with column ID. Specifications:- Endcapped: Yes. Guard Co lumn: No. Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab Poroshell 120. LC Column 1 * 1 Pce
N° de catalogue HEWL699775-702T
Fournisseur:  Agilent
Frequently Bought Together