ContactUsLinkComponent
InfinityLab Poroshell 120 HILIC, 2.1 x 50 mm, 4 um, narrow bore LC column, with column ID. Specifications:- Carbon Load: 0 %. Endcap ped: No. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab Poroshel 1 * 1 Pce
N° de catalogue HEWL699770-901T
Fournisseur:  Agilent
InfinityLab Poroshell 120 HILIC, 2.1 x 50 mm, 4 um, narrow bore LC column, with column ID. Specifications:- Carbon Load: 0 %. Endcap ped: No. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab Poroshel 1 * 1 Pce
N° de catalogue HEWL699770-901T
Fournisseur:  Agilent
Frequently Bought Together