InfinityLab Poroshell HPH-C18, 2.1 x 50 mm, 4 um, narrow bore LC column, with column ID. Specifications:- Endcapped: Yes. Guard Colu mn: No. Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab Poroshell 120. LC Column Ma 1 * 1 Pce
HEWL699770-702T
: Agilent
Frequently Bought Together