ContactUsLinkComponent
InfinityLab Poroshell 120 PFP, 2.1 x 50 mm, 1.9 um, narrow bore LC column, with column ID. Specifications:- Carbon Load: 5.1 %. Endc apped: Yes. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab Poros 1 * 1 Pce
Catalog # HEWL699675-408
Supplier:  Agilent
undefined
InfinityLab Poroshell 120 PFP, 2.1 x 50 mm, 1.9 um, narrow bore LC column, with column ID. Specifications:- Carbon Load: 5.1 %. Endc apped: Yes. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab Poros 1 * 1 Pce
Catalog # HEWL699675-408
Supplier:  Agilent
Frequently Bought Together