ContactUsLinkComponent
InfinityLab Poroshell 120 EC-C18, 3, 75 mm, 2.7 um, solvent saver LC column, with column ID. Specifications:- Carbon Load: 10 %. End capped: Yes. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 3 mm. LC Column Brand: InfinityLab Porosh 1 * 1 Pce
N° de catalogue HEWL697975-302T
Fournisseur:  Agilent
InfinityLab Poroshell 120 EC-C18, 3, 75 mm, 2.7 um, solvent saver LC column, with column ID. Specifications:- Carbon Load: 10 %. End capped: Yes. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 3 mm. LC Column Brand: InfinityLab Porosh 1 * 1 Pce
N° de catalogue HEWL697975-302T
Fournisseur:  Agilent
Frequently Bought Together