InfinityLab Poroshell 120 HILIC, 3.0 x 100 mm, 4 um, solvent saver LC column, with column ID. Specifications:- Carbon Load: 0 %. End capped: No. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 3 mm. LC Column Brand: InfinityLab Poroshe 1 * 1 Pce
N° de catalogue HEWL695970-301T
Fournisseur: Agilent
Frequently Bought Together